The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175063.5 | NP_778233.4 | 4 - 5 | 475 | 91 |
NM_206538.3 | NP_996261.1 | 4 - 5 | 475 | 91 | |
XM_006723163.3 | XP_006723226.1 | 4 - 5 | 469 | 91 | |
XM_011526818.2 | XP_011525120.1 | 4 - 5 | 469 | 91 | |
GenBank mRNA | AY194293.2 | - | 8 - 9 | 1433 | 91 |
AY358710.1 | - | 4 - 5 | 435 | 91 | |
AY761095.1 | - | 4 - 5 | 453 | 91 | |
AY761097.1 | - | 4 - 5 | 453 | 91 | |
BC032948.2 | - | 4 - 5 | 416 | 91 | |
BC035001.2 | - | 4 - 5 | 414 | 91 | |
BC062607.1 | - | 4 - 5 | 617 | 91 | |
CN423491.1 | - | 1 - 2 | 100 | 91 | |
HY149334.1 | - | 1 - 2 | 109 | 91 |