The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006638.2 | NP_001006639.1 | 5 - 6 | 421 | 77 |
NM_001163989.1 | NP_001157461.1 | 5 - 6 | 570 | 77 | |
NM_001163990.1 | NP_001157462.1 | 4 - 5 | 310 | 77 | |
NM_175738.4 | NP_783865.1 | 5 - 6 | 814 | 77 | |
XM_005257288.4 | XP_005257345.1 | 5 - 6 | 439 | 77 | |
GenBank mRNA | AK054846.1 | - | 5 - 6 | 751 | 77 |
AK098068.1 | - | 5 - 6 | 421 | 77 | |
AK127073.1 | - | 5 - 6 | 1353 | 77 | |
AK290202.1 | - | 5 - 6 | 790 | 77 | |
AK296062.1 | - | 5 - 6 | 409 | 77 | |
AK296172.1 | - | 4 - 5 | 273 | 77 | |
AK298033.1 | - | 4 - 5 | 306 | 77 | |
AK303442.1 | - | 5 - 6 | 473 | 77 | |
BC016615.1 | - | 5 - 6 | 367 | 77 | |
BC040547.1 | - | 5 - 6 | 800 | 77 |