The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193451.1 | NP_001180380.1 | 2 - 3 | 542 | 78 |
NM_175861.3 | NP_787057.2 | 2 - 3 | 633 | 78 | |
XM_005253499.4 | XP_005253556.1 | 2 - 3 | 608 | 78 | |
XM_017020003.1 | XP_016875492.1 | 2 - 3 | 609 | 78 | |
XM_017020004.1 | XP_016875493.1 | 2 - 3 | 608 | 78 | |
XM_017020005.1 | XP_016875494.1 | 2 - 3 | 607 | 78 | |
XM_017020006.1 | XP_016875495.1 | 2 - 3 | 612 | 78 | |
GenBank mRNA | AK127297.1 | - | 2 - 3 | 418 | 78 |
BC042083.1 | - | 2 - 3 | 633 | 78 | |
GQ865601.1 | - | 2 - 3 | 542 | 78 |