The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193451.1 | NP_001180380.1 | 11 - 12 | 1843 | 103 |
NM_175861.3 | NP_787057.2 | 11 - 12 | 1934 | 103 | |
XM_005253499.4 | XP_005253556.1 | 13 - 14 | 2095 | 103 | |
XM_017020003.1 | XP_016875492.1 | 12 - 13 | 1928 | 103 | |
XM_017020004.1 | XP_016875493.1 | 12 - 13 | 1888 | 103 | |
XM_017020005.1 | XP_016875494.1 | 10 - 11 | 1701 | 103 | |
XM_017020007.1 | XP_016875496.1 | 9 - 10 | 1121 | 103 | |
GenBank mRNA | AK055962.1 | - | 9 - 10 | 1095 | 103 |
AK123619.1 | - | 7 - 8 | 1525 | 103 | |
AL832328.1 | - | 2 - 3 | 259 | 103 | |
BC042083.1 | - | 11 - 12 | 1934 | 103 | |
GQ865601.1 | - | 11 - 12 | 1843 | 103 |