Product Details

Assay Reference Genome
Location

Chr.19:44790021 on build GRCh38
Cytoband
19q13.32
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
23624
Gene Symbol
CBLC
Gene Name
Cbl proto-oncogene C
Gene Aliases
CBL-3, CBL-SL, RNF57
Location
Chr.19:44777855-44800646 on build GRCh38
Assay Gene Location
Overlaps Intron 5 - Intron 6
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
CBLC NM_001130852.1 NP_001124324.1
NM_012116.3 NP_036248.3
XM_005258696.3 XP_005258753.1
XM_011526688.2 XP_011524990.1
XM_011526689.2 XP_011524991.1
XM_011526690.2 XP_011524992.1
AB028645.1 BAA86298.1
AF117646.1 AAD34341.1
AF117647.1 AAD34342.1
BC028915.1 AAH28915.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv833841 Chr19:44705217 - 44804786 on Build GRCh38 Loss CEACAM16 MIR8085 CBLC BCL3
nsv2506 Chr19:44750006 - 44794554 on Build GRCh38 Deletion MIR8085 CBLC BCL3

View Full Product Details