Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRPM5
Entrez Gene ID
29850
Gene Name
transient receptor potential cation channel subfamily M member 5
Gene Aliases
LTRPC5, MTR1
Chromosome Location
Chr.11: 2403962 - 2423424 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014555.3 NP_055370.1 6 - 7 920 72
XM_017017628.1 XP_016873117.1 7 - 8 1007 72
GenBank mRNA AF177473.1 - 6 - 7 920 72
AJ270996.1 - 6 - 7 893 72
BC093787.1 - 6 - 7 917 72
BC093789.1 - 6 - 7 917 72
BC143351.1 - 6 - 7 917 72
BC143353.1 - 6 - 7 917 72

View Full Product Details