Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRPC7
Entrez Gene ID
57113
Gene Name
transient receptor potential cation channel subfamily C member 7
Gene Aliases
TRP7
Chromosome Location
Chr.5: 136213311 - 136365476 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167576.1 NP_001161048.1 6 - 7 1916 93
NM_001167577.1 NP_001161049.1 7 - 8 2081 93
NM_020389.2 NP_065122.1 8 - 9 2264 93
GenBank mRNA AJ272034.1 - 7 - 8 2042 93
AJ421783.1 - 7 - 8 2042 93
AJ549088.1 - 6 - 7 1859 93
AJ549089.1 - 6 - 7 1877 93
AJ549090.1 - 4 - 5 1477 93
AY167926.1 - 6 - 7 1874 93
AY167927.1 - 5 - 6 1694 93
BC128185.1 - 7 - 8 2042 93
BC128186.1 - 7 - 8 2058 93

View Full Product Details