Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRPC7
Entrez Gene ID
57113
Gene Name
transient receptor potential cation channel subfamily C member 7
Gene Aliases
TRP7
Chromosome Location
Chr.5: 136213311 - 136365476 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167576.1 NP_001161048.1 7 - 8 2135 103
NM_001167577.1 NP_001161049.1 8 - 9 2300 103
NM_020389.2 NP_065122.1 9 - 10 2483 103
GenBank mRNA AJ272034.1 - 8 - 9 2261 103
AJ421783.1 - 8 - 9 2261 103
AJ549088.1 - 7 - 8 2078 103
AJ549089.1 - 7 - 8 2096 103
AJ549090.1 - 5 - 6 1696 103
AY167926.1 - 7 - 8 2093 103
AY167927.1 - 6 - 7 1913 103
BC128185.1 - 8 - 9 2261 103
BC128186.1 - 8 - 9 2277 103
BI826927.1 - 2 - 3 260 103

View Full Product Details