Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2E1
Entrez Gene ID
1571
Gene Name
cytochrome P450 family 2 subfamily E member 1
Gene Aliases
CPE1, CYP2E, P450-J, P450C2E
Chromosome Location
Chr.10: 133527363 - 133539116 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000773.3 NP_000764.1 6 - 7 1002 74
GenBank mRNA AF084225.1 - 6 - 7 876 74
AF182276.1 - 6 - 7 977 74
AJ853939.1 - 5 - 6 708 74
AJ853940.1 - 4 - 5 558 74
AK290822.1 - 6 - 7 1002 74
BC067433.1 - 6 - 7 972 74
BC067435.1 - 6 - 7 973 74
DQ149222.1 - 6 - 7 986 74
J02625.1 - 6 - 7 981 74

View Full Product Details