Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PC
Entrez Gene ID
5091
Gene Name
pyruvate carboxylase
Gene Aliases
PCB
Chromosome Location
Chr.11: 66848522 - 66958418 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000920.3 NP_000911.2 9 - 10 1113 61
NM_001040716.1 NP_001035806.1 10 - 11 1301 61
NM_022172.2 NP_071504.2 8 - 9 1068 61
XM_005274031.4 XP_005274088.1 8 - 9 1160 61
XM_005274032.4 XP_005274089.1 8 - 9 1042 61
XM_006718578.3 XP_006718641.1 8 - 9 1113 61
XM_011545086.2 XP_011543388.1 8 - 9 1429 61
XM_017017868.1 XP_016873357.1 11 - 12 1600 61
XM_017017869.1 XP_016873358.1 11 - 12 1471 61
XM_017017870.1 XP_016873359.1 10 - 11 1412 61
XM_017017871.1 XP_016873360.1 10 - 11 1283 61
XM_017017872.1 XP_016873361.1 9 - 10 1665 61
GenBank mRNA AK297705.1 - 9 - 10 1113 61
BC011617.2 - 9 - 10 1102 61
S72370.1 - 8 - 9 1101 61
U04641.1 - 8 - 9 1057 61
U30891.1 - 9 - 10 1071 61

View Full Product Details