The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001156.4 | NP_001147.1 | 8 - 9 | 819 | 136 |
NM_001320879.1 | NP_001307808.1 | 7 - 8 | 764 | 136 | |
NM_001320880.1 | NP_001307809.1 | 8 - 9 | 830 | 136 | |
NM_004034.3 | NP_004025.1 | 9 - 10 | 885 | 136 | |
XM_017016162.1 | XP_016871651.1 | 9 - 10 | 921 | 136 | |
XM_017016163.1 | XP_016871652.1 | 8 - 9 | 855 | 136 | |
GenBank mRNA | AB062429.1 | - | 8 - 9 | 777 | 136 |
AK295319.1 | - | 8 - 9 | 696 | 136 | |
AK300086.1 | - | 6 - 7 | 586 | 136 | |
AK301681.1 | - | 8 - 9 | 573 | 136 | |
AK309141.1 | - | 8 - 9 | 581 | 136 | |
AK312447.1 | - | 9 - 10 | 858 | 136 | |
AK313024.1 | - | 8 - 9 | 762 | 136 | |
BC001322.1 | - | 1 - 2 | 70 | 136 | |
BC002632.2 | - | 8 - 9 | 779 | 136 | |
BT007187.1 | - | 7 - 8 | 744 | 136 | |
BX361177.2 | - | 7 - 8 | 730 | 136 | |
CR407686.1 | - | 7 - 8 | 744 | 136 | |
J04543.1 | - | 8 - 9 | 804 | 136 |