The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002208.4 | NP_002199.3 | 21 - 22 | 2755 | 71 |
XM_011523823.2 | XP_011522125.1 | 21 - 22 | 3193 | 71 | |
XM_011523825.2 | XP_011522127.1 | 21 - 22 | 3191 | 71 | |
XM_017024586.1 | XP_016880075.1 | 22 - 23 | 4578 | 71 | |
GenBank mRNA | BC113436.1 | - | 21 - 22 | 2745 | 71 |
BC117207.1 | - | 21 - 22 | 2745 | 71 | |
L25851.2 | - | 21 - 22 | 2781 | 71 |