The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005999.2 | NP_005990.1 | 4 - 5 | 531 | 68 |
GenBank mRNA | AK313068.1 | - | 4 - 5 | 469 | 68 |
BC010376.1 | - | 4 - 5 | 489 | 68 | |
BC011797.1 | - | 4 - 5 | 506 | 68 | |
BX537610.1 | - | 2 - 3 | 3334 | 68 | |
X95073.1 | - | 4 - 5 | 533 | 68 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_028394.1 | 4 - 5 | 531 | 68 | |
NR_028395.1 | 4 - 5 | 531 | 68 | ||
NR_028397.1 | 4 - 5 | 531 | 68 | ||
NR_028399.1 | 4 - 5 | 531 | 68 |