Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KERA
Entrez Gene ID
11081
Gene Name
keratocan
Gene Aliases
CNA2, KTN, SLRR2B
Chromosome Location
Chr.12: 91050491 - 91058354 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007035.3 NP_008966.1 1 - 2 607 64
GenBank mRNA AF205403.1 - 1 - 2 236 64
AK314990.1 - 1 - 2 169 64
BM722433.1 - 1 - 2 140 64
CV571245.1 - 1 - 2 154 64

View Full Product Details