The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317960.1 | NP_001304889.1 | 10 - 11 | 1205 | 92 |
NM_012255.4 | NP_036387.2 | 10 - 11 | 1045 | 92 | |
XM_017027722.1 | XP_016883211.1 | 10 - 11 | 1200 | 92 | |
XM_017027723.1 | XP_016883212.1 | 10 - 11 | 974 | 92 | |
XM_017027724.1 | XP_016883213.1 | 10 - 11 | 1200 | 92 | |
GenBank mRNA | AF064257.1 | - | 10 - 11 | 998 | 92 |
AK001573.1 | - | 10 - 11 | 1015 | 92 | |
AK302846.1 | - | 10 - 11 | 909 | 92 | |
AK303312.1 | - | 9 - 10 | 856 | 92 | |
AL136841.1 | - | 10 - 11 | 971 | 92 | |
AY382900.1 | - | 9 - 10 | 860 | 92 |