The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317960.1 | NP_001304889.1 | 17 - 18 | 1938 | 97 |
NM_012255.4 | NP_036387.2 | 17 - 18 | 1778 | 97 | |
XM_017027723.1 | XP_016883212.1 | 17 - 18 | 1731 | 97 | |
GenBank mRNA | AF064257.1 | - | 17 - 18 | 1731 | 97 |
AF152169.1 | - | 17 - 18 | 1681 | 97 | |
AK000084.1 | - | 6 - 7 | 593 | 97 | |
AK001573.1 | - | 17 - 18 | 1748 | 97 | |
AK302846.1 | - | 17 - 18 | 1642 | 97 | |
AK303312.1 | - | 16 - 17 | 1589 | 97 | |
AK314602.1 | - | 17 - 18 | 1717 | 97 | |
AL136841.1 | - | 17 - 18 | 1704 | 97 | |
AY382900.1 | - | 16 - 17 | 1593 | 97 | |
BC006417.1 | - | 5 - 6 | 553 | 97 |