The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317960.1 | NP_001304889.1 | 5 - 6 | 762 | 126 |
NM_012255.4 | NP_036387.2 | 5 - 6 | 602 | 126 | |
XM_017027722.1 | XP_016883211.1 | 5 - 6 | 757 | 126 | |
XM_017027723.1 | XP_016883212.1 | 5 - 6 | 531 | 126 | |
XM_017027724.1 | XP_016883213.1 | 5 - 6 | 757 | 126 | |
GenBank mRNA | AF064257.1 | - | 5 - 6 | 555 | 126 |
AF152169.1 | - | 5 - 6 | 505 | 126 | |
AK001573.1 | - | 5 - 6 | 572 | 126 | |
AK302846.1 | - | 5 - 6 | 466 | 126 | |
AK303312.1 | - | 4 - 5 | 413 | 126 | |
AK314602.1 | - | 5 - 6 | 541 | 126 | |
AL136841.1 | - | 5 - 6 | 528 | 126 | |
AY382900.1 | - | 4 - 5 | 417 | 126 | |
CF994999.1 | - | 5 - 6 | 609 | 126 |