The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012430.4 | NP_036562.2 | 4 - 5 | 623 | 67 |
XM_011512673.2 | XP_011510975.1 | 4 - 5 | 2139 | 67 | |
XM_011512675.2 | XP_011510977.1 | 5 - 6 | 790 | 67 | |
XM_011512676.2 | XP_011510978.1 | 4 - 5 | 679 | 67 | |
GenBank mRNA | AF100749.1 | - | 4 - 5 | 605 | 67 |
AK057587.1 | - | 3 - 4 | 1439 | 67 | |
AK315771.1 | - | 4 - 5 | 592 | 67 | |
BC007122.1 | - | 3 - 4 | 571 | 67 |