The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012448.3 | NP_036580.2 | 11 - 12 | 1555 | 73 |
XM_005257626.4 | XP_005257683.1 | 11 - 12 | 1609 | 73 | |
XM_017024977.1 | XP_016880466.1 | 10 - 11 | 1374 | 73 | |
GenBank mRNA | AB208920.1 | - | 11 - 12 | 1534 | 73 |
AK308163.1 | - | 11 - 12 | 1443 | 73 | |
BC065227.1 | - | 11 - 12 | 1555 | 73 | |
U47686.1 | - | 11 - 12 | 1532 | 73 | |
U48730.2 | - | 10 - 11 | 1397 | 73 |