The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012448.3 | NP_036580.2 | 2 - 3 | 298 | 91 |
XM_005257626.4 | XP_005257683.1 | 2 - 3 | 352 | 91 | |
GenBank mRNA | AB208920.1 | - | 2 - 3 | 277 | 91 |
AK308163.1 | - | 2 - 3 | 186 | 91 | |
BC020868.1 | - | 2 - 3 | 298 | 91 | |
BC065227.1 | - | 2 - 3 | 298 | 91 | |
U47686.1 | - | 2 - 3 | 275 | 91 | |
U48730.2 | - | 1 - 2 | 140 | 91 |