Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PNKD
Entrez Gene ID
25953
Gene Name
paroxysmal nonkinesigenic dyskinesia
Gene Aliases
BRP17, DYT8, FKSG19, FPD1, KIPP1184, MR-1, MR1, PDC, PKND1, TAHCCP2
Chromosome Location
Chr.2: 218270392 - 218346793 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077399.2 NP_001070867.1 1 - 2 214 106
NM_015488.4 NP_056303.3 1 - 2 214 106
XM_017003771.1 XP_016859260.1 1 - 2 121 106
GenBank mRNA AF318057.1 - 1 - 2 214 106
AF417001.1 - 1 - 2 214 106
AK289867.1 - 1 - 2 122 106
AL080092.1 - 1 - 2 96 106
AY039043.1 - - 102 106
BC021118.1 - 1 - 2 101 106

View Full Product Details