The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020139.3 | NP_064524.3 | 7 - 8 | 635 | 85 |
XM_006714274.3 | XP_006714337.1 | 8 - 9 | 672 | 85 | |
XM_011532127.2 | XP_011530429.1 | 8 - 9 | 714 | 85 | |
XM_011532128.2 | XP_011530430.1 | 7 - 8 | 672 | 85 | |
XM_017008461.1 | XP_016863950.1 | 8 - 9 | 711 | 85 | |
XM_017008462.1 | XP_016863951.1 | 7 - 8 | 669 | 85 | |
GenBank mRNA | AF164790.1 | - | 7 - 8 | 599 | 85 |
AK023323.1 | - | 6 - 7 | 1608 | 85 | |
AK128463.1 | - | 7 - 8 | 2350 | 85 | |
AK290160.1 | - | 7 - 8 | 591 | 85 | |
AK300626.1 | - | 7 - 8 | 480 | 85 | |
AY358841.1 | - | 7 - 8 | 601 | 85 | |
BC001953.1 | - | 7 - 8 | 628 | 85 | |
BC037277.2 | - | 6 - 7 | 1614 | 85 | |
BC095414.1 | - | 7 - 8 | 603 | 85 |