Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTMR14
Entrez Gene ID
64419
Gene Name
myotubularin related protein 14
Gene Aliases
C3orf29
Chromosome Location
Chr.3: 9649433 - 9702394 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077525.2 NP_001070993.1 5 - 6 702 103
NM_001077526.2 NP_001070994.1 5 - 6 702 103
NM_022485.4 NP_071930.2 5 - 6 702 103
XM_005265400.1 XP_005265457.1 5 - 6 675 103
XM_011534012.1 XP_011532314.1 4 - 5 471 103
XM_011534013.1 XP_011532315.1 4 - 5 427 103
XM_017007034.1 XP_016862523.1 6 - 7 872 103
XM_017007035.1 XP_016862524.1 6 - 7 872 103
XM_017007036.1 XP_016862525.1 6 - 7 871 103
XM_017007037.1 XP_016862526.1 6 - 7 872 103
XM_017007038.1 XP_016862527.1 5 - 6 675 103
XM_017007039.1 XP_016862528.1 6 - 7 871 103
XM_017007040.1 XP_016862529.1 6 - 7 872 103
XM_017007041.1 XP_016862530.1 4 - 5 471 103
XM_017007042.1 XP_016862531.1 5 - 6 675 103
XM_017007043.1 XP_016862532.1 4 - 5 471 103
XM_017007044.1 XP_016862533.1 4 - 5 593 103
GenBank mRNA AF289603.1 - 5 - 6 665 103
AK026058.1 - 4 - 5 269 103
AK074792.1 - 5 - 6 673 103
AK295891.1 - 4 - 5 554 103
AL713695.1 - 5 - 6 623 103
AY007098.1 - 2 - 3 110 103
BC001674.1 - 5 - 6 607 103
BC025952.1 - 5 - 6 652 103
BC035690.1 - 5 - 6 614 103
BC050626.1 - 5 - 6 625 103
DQ630520.1 - 5 - 6 551 103

View Full Product Details