The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317062.1 | NP_001303991.1 | 9 - 10 | 1046 | 66 |
NM_001317064.1 | NP_001303993.1 | 7 - 8 | 818 | 66 | |
NM_024102.3 | NP_077007.1 | 9 - 10 | 1010 | 66 | |
NR_133654.1 | 8 - 9 | 868 | 66 | ||
GenBank mRNA | AB073603.1 | - | 9 - 10 | 958 | 66 |
AB074171.1 | - | 4 - 5 | 322 | 66 | |
AF052183.1 | - | 9 - 10 | 896 | 66 | |
AF478464.1 | - | 9 - 10 | 871 | 66 | |
AK022860.1 | - | 9 - 10 | 940 | 66 | |
AK223189.1 | - | 9 - 10 | 940 | 66 | |
AK298179.1 | - | 7 - 8 | 762 | 66 | |
AY225316.1 | - | 9 - 10 | 896 | 66 | |
BC001679.2 | - | 9 - 10 | 931 | 66 | |
BC006477.1 | - | 9 - 10 | 931 | 66 | |
BC009411.1 | - | 9 - 10 | 943 | 66 | |
BC011778.2 | - | 9 - 10 | 950 | 66 | |
BC016946.1 | - | 9 - 10 | 910 | 66 |