Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTYH2
Entrez Gene ID
94015
Gene Name
tweety family member 2
Gene Aliases
C17orf29
Chromosome Location
Chr.17: 74213557 - 74262020 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032646.5 NP_116035.5 10 - 11 1147 93
NM_052869.1 NP_443101.1 4 - 5 957 93
XM_005257824.3 XP_005257881.1 10 - 11 1075 93
GenBank mRNA AF319952.1 - 10 - 11 1126 93
AK091085.1 - 8 - 9 828 93
AK126955.1 - 4 - 5 1365 93
AK296511.1 - 10 - 11 1070 93
AK315163.1 - 10 - 11 1133 93
BC005168.1 - 10 - 11 1122 93
BC107492.1 - 10 - 11 1143 93
BC107602.1 - 10 - 11 1143 93

View Full Product Details