Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTYH2
Entrez Gene ID
94015
Gene Name
tweety family member 2
Gene Aliases
C17orf29
Chromosome Location
Chr.17: 74213557 - 74262020 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032646.5 NP_116035.5 1 - 2 165 49
GenBank mRNA AF319952.1 - 1 - 2 144 49
AK315163.1 - 1 - 2 151 49
BC005168.1 - 1 - 2 140 49
BC107492.1 - 1 - 2 161 49
BC107602.1 - 1 - 2 161 49

View Full Product Details