Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68214193-68214420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLN6
Entrez Gene ID
54982
Gene Name
ceroid-lipofuscinosis, neuronal 6, late infantile, variant
Gene Aliases
CLN4A, HsT18960, nclf
UniGene
Hs.584921
Gene Chromosome Location
Chr.15: 68206992-68229742 on Build GRCh38
Transcripts
NM_017882.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
111530 c.251delA p.Y84fs*32 Chr.15: 68214336-68214336 on Build GRCh38
6630567 c.269A>C p.N90T Chr.15: 68214318-68214318 on Build GRCh38
6332242 c.281C>T p.P94L Chr.15: 68214306-68214306 on Build GRCh38
6630565 c.271G>A p.V91I Chr.15: 68214316-68214316 on Build GRCh38
298071 c.203T>C p.V68A Chr.15: 68214384-68214384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778856506 Chr.15: 68214268-68214268 on Build GRCh38
rs756256935 Chr.15: 68214257-68214257 on Build GRCh38
rs371705916 Chr.15: 68214393-68214393 on Build GRCh38
rs759935979 Chr.15: 68214388-68214388 on Build GRCh38
rs767945732 Chr.15: 68214392-68214392 on Build GRCh38
rs371130165 Chr.15: 68214255-68214255 on Build GRCh38
rs781532461 Chr.15: 68214252-68214252 on Build GRCh38
rs776974644 Chr.15: 68214357-68214357 on Build GRCh38
rs909178698 Chr.15: 68214343-68214343 on Build GRCh38
rs753011260 Chr.15: 68214339-68214339 on Build GRCh38
rs573090657 Chr.15: 68214251-68214251 on Build GRCh38
rs1030759897 Chr.15: 68214286-68214286 on Build GRCh38
rs545955828 Chr.15: 68214332-68214332 on Build GRCh38
rs745619273 Chr.15: 68214272-68214272 on Build GRCh38
rs1023750698 Chr.15: 68214241-68214241 on Build GRCh38
rs752943329 Chr.15: 68214263-68214264 on Build GRCh38
rs747711900 Chr.15: 68214378-68214378 on Build GRCh38
rs146782642 Chr.15: 68214291-68214291 on Build GRCh38
rs777976727 Chr.15: 68214260-68214260 on Build GRCh38
rs154774641 Chr.15: 68214356-68214356 on Build GRCh38
rs769286184 Chr.15: 68214379-68214379 on Build GRCh38
rs786205067 Chr.15: 68214315-68214316 on Build GRCh38
rs544291935 Chr.15: 68214362-68214362 on Build GRCh38
rs146980624 Chr.15: 68214374-68214374 on Build GRCh38
rs1040306361 Chr.15: 68214242-68214242 on Build GRCh38
rs61224699 Chr.15: 68214275-68214275 on Build GRCh38
rs104894483 Chr.15: 68214373-68214373 on Build GRCh38
rs979566677 Chr.15: 68214284-68214284 on Build GRCh38
rs764301084 Chr.15: 68214350-68214350 on Build GRCh38
rs752816745 Chr.15: 68214340-68214340 on Build GRCh38
rs746613593 Chr.15: 68214308-68214308 on Build GRCh38
rs766809818 Chr.15: 68214321-68214321 on Build GRCh38
rs774960579 Chr.15: 68214383-68214383 on Build GRCh38
rs756241175 Chr.15: 68214348-68214348 on Build GRCh38
rs575895650 Chr.15: 68214271-68214271 on Build GRCh38
rs749259550 Chr.15: 68214265-68214265 on Build GRCh38
rs940519581 Chr.15: 68214234-68214234 on Build GRCh38
rs767741029 Chr.15: 68214336-68214336 on Build GRCh38
rs140519790 Chr.15: 68214316-68214316 on Build GRCh38
rs150001589 Chr.15: 68214309-68214309 on Build GRCh38
rs532180601 Chr.15: 68214328-68214328 on Build GRCh38
rs796052351 Chr.15: 68214289-68214289 on Build GRCh38
rs967752328 Chr.15: 68214273-68214273 on Build GRCh38
rs145247814 Chr.15: 68214317-68214317 on Build GRCh38
rs139261571 Chr.15: 68214305-68214305 on Build GRCh38
rs154774633 Chr.15: 68214387-68214387 on Build GRCh38
rs199658226 Chr.15: 68214366-68214366 on Build GRCh38
rs796052350 Chr.15: 68214302-68214302 on Build GRCh38
rs771752231 Chr.15: 68214276-68214276 on Build GRCh38
rs956493858 Chr.15: 68214287-68214287 on Build GRCh38

View Full Product Details