Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40395556-40395829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRX
Entrez Gene ID
57716
Gene Name
periaxin
Gene Aliases
CMT4F
UniGene
Hs.205457
Gene Chromosome Location
Chr.19: 40393762-40414718 on Build GRCh38
Transcripts
NM_020956.2 NM_181882.2 XM_011527171.2 XM_017027046.1 XM_017027047.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1723816 c.2615_2616insC p.A873fs*3 Chr.19: 40395736-40395737 on Build GRCh38
6427020 c.2623A>T p.K875* Chr.19: 40395729-40395729 on Build GRCh38
474749 c.2731G>A p.V911M Chr.19: 40395621-40395621 on Build GRCh38
3534060 c.2654C>T p.P885L Chr.19: 40395698-40395698 on Build GRCh38
4140646 c.2775C>G p.V925V Chr.19: 40395577-40395577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61735546 Chr.19: 40395625-40395625 on Build GRCh38
rs141686828 Chr.19: 40395804-40395804 on Build GRCh38
rs570322417 Chr.19: 40395809-40395809 on Build GRCh38
rs1033917570 Chr.19: 40395683-40395683 on Build GRCh38
rs537099688 Chr.19: 40395763-40395763 on Build GRCh38
rs775808575 Chr.19: 40395796-40395796 on Build GRCh38
rs148127181 Chr.19: 40395759-40395759 on Build GRCh38
rs761066015 Chr.19: 40395733-40395733 on Build GRCh38
rs374001049 Chr.19: 40395736-40395737 on Build GRCh38
rs766388687 Chr.19: 40395635-40395635 on Build GRCh38
rs377457671 Chr.19: 40395621-40395621 on Build GRCh38
rs772208597 Chr.19: 40395792-40395792 on Build GRCh38
rs769587753 Chr.19: 40395705-40395705 on Build GRCh38
rs747871484 Chr.19: 40395703-40395703 on Build GRCh38
rs375201649 Chr.19: 40395711-40395711 on Build GRCh38
rs201270794 Chr.19: 40395675-40395675 on Build GRCh38
rs779594126 Chr.19: 40395743-40395743 on Build GRCh38
rs773084101 Chr.19: 40395706-40395706 on Build GRCh38
rs139005780 Chr.19: 40395701-40395701 on Build GRCh38
rs752531623 Chr.19: 40395662-40395662 on Build GRCh38
rs1017754371 Chr.19: 40395779-40395779 on Build GRCh38
rs373698395 Chr.19: 40395586-40395586 on Build GRCh38
rs144639467 Chr.19: 40395603-40395603 on Build GRCh38
rs756689732 Chr.19: 40395663-40395663 on Build GRCh38
rs268671 Chr.19: 40395707-40395707 on Build GRCh38
rs771736913 Chr.19: 40395740-40395743 on Build GRCh38
rs754359052 Chr.19: 40395741-40395741 on Build GRCh38
rs759471716 Chr.19: 40395773-40395773 on Build GRCh38
rs767702501 Chr.19: 40395715-40395715 on Build GRCh38
rs932643968 Chr.19: 40395616-40395616 on Build GRCh38
rs878855283 Chr.19: 40395622-40395622 on Build GRCh38
rs778360076 Chr.19: 40395671-40395671 on Build GRCh38
rs754521128 Chr.19: 40395745-40395745 on Build GRCh38
rs745442249 Chr.19: 40395602-40395602 on Build GRCh38
rs201389706 Chr.19: 40395740-40395740 on Build GRCh38
rs768242642 Chr.19: 40395700-40395700 on Build GRCh38
rs145790961 Chr.19: 40395732-40395732 on Build GRCh38
rs751096431 Chr.19: 40395744-40395744 on Build GRCh38
rs777582529 Chr.19: 40395761-40395761 on Build GRCh38
rs778762350 Chr.19: 40395655-40395655 on Build GRCh38
rs770828114 Chr.19: 40395768-40395768 on Build GRCh38
rs188765166 Chr.19: 40395637-40395637 on Build GRCh38
rs531650268 Chr.19: 40395598-40395598 on Build GRCh38
rs148451350 Chr.19: 40395717-40395717 on Build GRCh38
rs268672 Chr.19: 40395697-40395697 on Build GRCh38
rs781036230 Chr.19: 40395702-40395702 on Build GRCh38
rs767898499 Chr.19: 40395651-40395651 on Build GRCh38
rs775152960 Chr.19: 40395606-40395606 on Build GRCh38
rs747611612 Chr.19: 40395648-40395649 on Build GRCh38
rs778721394 Chr.19: 40395588-40395589 on Build GRCh38
rs268673 Chr.19: 40395589-40395589 on Build GRCh38
rs570264094 Chr.19: 40395710-40395710 on Build GRCh38
rs145203783 Chr.19: 40395624-40395624 on Build GRCh38
rs757934238 Chr.19: 40395681-40395681 on Build GRCh38
rs941768924 Chr.19: 40395645-40395645 on Build GRCh38
rs201792838 Chr.19: 40395577-40395577 on Build GRCh38
rs774488422 Chr.19: 40395769-40395769 on Build GRCh38
rs561046470 Chr.19: 40395590-40395590 on Build GRCh38
rs781439409 Chr.19: 40395578-40395578 on Build GRCh38
rs780763165 Chr.19: 40395750-40395750 on Build GRCh38
rs779463585 Chr.19: 40395696-40395696 on Build GRCh38
rs746664787 Chr.19: 40395610-40395610 on Build GRCh38
rs899773763 Chr.19: 40395608-40395608 on Build GRCh38
rs747820251 Chr.19: 40395754-40395754 on Build GRCh38

View Full Product Details