Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 107639420-107639665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPS1
Entrez Gene ID
5631
Gene Name
phosphoribosyl pyrophosphate synthetase 1
Gene Aliases
ARTS, CMTX5, DFN2, DFNX1, PPRibP, PRS-I, PRSI
UniGene
Hs.56
Gene Chromosome Location
Chr.X: 107628424-107651026 on Build GRCh38
Transcripts
NM_001204402.1 NM_002764.3

SNP Detail

SNP ID Chromosome Location
rs902859863 Chr.X: 107639599-107639599 on Build GRCh38
rs1033217069 Chr.X: 107639619-107639619 on Build GRCh38
rs766786656 Chr.X: 107639442-107639442 on Build GRCh38
rs752194539 Chr.X: 107639460-107639460 on Build GRCh38
rs1015038412 Chr.X: 107639563-107639563 on Build GRCh38
rs755093762 Chr.X: 107639478-107639479 on Build GRCh38
rs998598761 Chr.X: 107639616-107639616 on Build GRCh38
rs11797233 Chr.X: 107639573-107639573 on Build GRCh38
rs1029293242 Chr.X: 107639554-107639554 on Build GRCh38
rs777885394 Chr.X: 107639495-107639496 on Build GRCh38
rs758657216 Chr.X: 107639479-107639480 on Build GRCh38

View Full Product Details