Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52315932-52316157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT83
Entrez Gene ID
3889
Gene Name
keratin 83
Gene Aliases
HB3, Hb-3, KRTHB3
UniGene
Hs.720768
Gene Chromosome Location
Chr.12: 52314301-52321398 on Build GRCh38
Transcripts
NM_002282.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
302522 c.1113C>A p.C371* Chr.12: 52316042-52316042 on Build GRCh38
6256137 c.1158C>A p.D386E Chr.12: 52315997-52315997 on Build GRCh38
2010336 c.1110C>T p.R370R Chr.12: 52316045-52316045 on Build GRCh38
147521 c.1056A>G p.E352E Chr.12: 52316099-52316099 on Build GRCh38
940672 c.1094C>A p.A365D Chr.12: 52316061-52316061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140635030 Chr.12: 52316071-52316071 on Build GRCh38
rs769731414 Chr.12: 52315971-52315971 on Build GRCh38
rs541107858 Chr.12: 52315961-52315961 on Build GRCh38
rs745551113 Chr.12: 52315996-52315996 on Build GRCh38
rs201054359 Chr.12: 52316020-52316020 on Build GRCh38
rs753918513 Chr.12: 52316126-52316126 on Build GRCh38
rs765209287 Chr.12: 52316093-52316093 on Build GRCh38
rs748566173 Chr.12: 52316061-52316061 on Build GRCh38
rs761585220 Chr.12: 52315998-52315998 on Build GRCh38
rs964369709 Chr.12: 52316033-52316033 on Build GRCh38
rs776130197 Chr.12: 52315987-52315987 on Build GRCh38
rs144732230 Chr.12: 52315992-52315992 on Build GRCh38
rs770479449 Chr.12: 52316075-52316075 on Build GRCh38
rs559477397 Chr.12: 52316054-52316054 on Build GRCh38
rs1001331106 Chr.12: 52316058-52316058 on Build GRCh38
rs200402665 Chr.12: 52316117-52316117 on Build GRCh38
rs372980511 Chr.12: 52316046-52316046 on Build GRCh38
rs571320078 Chr.12: 52316095-52316095 on Build GRCh38
rs749798524 Chr.12: 52315988-52315988 on Build GRCh38
rs148519033 Chr.12: 52316021-52316021 on Build GRCh38
rs985719651 Chr.12: 52316015-52316015 on Build GRCh38
rs747874210 Chr.12: 52315960-52315962 on Build GRCh38
rs755083762 Chr.12: 52316127-52316127 on Build GRCh38
rs756220670 Chr.12: 52316101-52316101 on Build GRCh38
rs2248473 Chr.12: 52316099-52316099 on Build GRCh38
rs200684354 Chr.12: 52316063-52316063 on Build GRCh38
rs765049913 Chr.12: 52316024-52316024 on Build GRCh38
rs772711995 Chr.12: 52316008-52316008 on Build GRCh38
rs151316742 Chr.12: 52315962-52315962 on Build GRCh38
rs759494546 Chr.12: 52316091-52316091 on Build GRCh38
rs779439957 Chr.12: 52316060-52316060 on Build GRCh38
rs773024879 Chr.12: 52316037-52316037 on Build GRCh38
rs2852468 Chr.12: 52316096-52316096 on Build GRCh38
rs1032488954 Chr.12: 52315974-52315974 on Build GRCh38
rs772979616 Chr.12: 52316070-52316070 on Build GRCh38
rs776078933 Chr.12: 52316090-52316090 on Build GRCh38
rs774787911 Chr.12: 52315995-52315995 on Build GRCh38
rs769672721 Chr.12: 52315976-52315984 on Build GRCh38
rs529158513 Chr.12: 52316032-52316032 on Build GRCh38
rs752598688 Chr.12: 52316094-52316094 on Build GRCh38
rs754844434 Chr.12: 52316022-52316022 on Build GRCh38
rs577607601 Chr.12: 52315975-52315975 on Build GRCh38
rs528339841 Chr.12: 52316055-52316055 on Build GRCh38
rs542589604 Chr.12: 52316047-52316047 on Build GRCh38
rs752441578 Chr.12: 52316026-52316026 on Build GRCh38
rs767145882 Chr.12: 52316000-52316000 on Build GRCh38
rs375064664 Chr.12: 52316052-52316052 on Build GRCh38

View Full Product Details