Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8584807-8585080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS10
Entrez Gene ID
81794
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 10
Gene Aliases
ADAM-TS10, ADAMTS-10, WMS, WMS1
UniGene
Hs.657508
Gene Chromosome Location
Chr.19: 8580240-8610735 on Build GRCh38
Transcripts
NM_001282352.1 NM_030957.3 XM_006722917.3 XM_017027338.1 XM_017027339.1 XM_017027340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7275092 c.3192C>T p.D1064D Chr.19: 8584905-8584905 on Build GRCh38
4542436 c.3188G>A p.G1063E Chr.19: 8584909-8584909 on Build GRCh38
4542106 c.3126G>A p.T1042T Chr.19: 8584971-8584971 on Build GRCh38
4820433 c.3136C>T p.R1046W Chr.19: 8584961-8584961 on Build GRCh38
7354242 c.3181A>G p.T1061A Chr.19: 8584916-8584916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547039905 Chr.19: 8584919-8584919 on Build GRCh38
rs782672084 Chr.19: 8584864-8584864 on Build GRCh38
rs1005172514 Chr.19: 8584873-8584873 on Build GRCh38
rs138501563 Chr.19: 8585059-8585060 on Build GRCh38
rs782617003 Chr.19: 8584910-8584910 on Build GRCh38
rs80046653 Chr.19: 8584904-8584904 on Build GRCh38
rs759098979 Chr.19: 8585062-8585062 on Build GRCh38
rs550993677 Chr.19: 8584874-8584874 on Build GRCh38
rs992529957 Chr.19: 8584925-8584925 on Build GRCh38
rs781916006 Chr.19: 8585016-8585016 on Build GRCh38
rs886054710 Chr.19: 8584882-8584882 on Build GRCh38
rs782376436 Chr.19: 8584952-8584952 on Build GRCh38
rs782317309 Chr.19: 8584909-8584909 on Build GRCh38
rs935826954 Chr.19: 8584996-8584996 on Build GRCh38
rs782096481 Chr.19: 8585059-8585059 on Build GRCh38
rs886054711 Chr.19: 8584938-8584938 on Build GRCh38
rs782258526 Chr.19: 8584856-8584856 on Build GRCh38
rs979468533 Chr.19: 8584986-8584986 on Build GRCh38
rs969473391 Chr.19: 8584959-8584959 on Build GRCh38
rs782323306 Chr.19: 8585006-8585006 on Build GRCh38
rs566999321 Chr.19: 8584981-8584981 on Build GRCh38
rs916677846 Chr.19: 8584948-8584948 on Build GRCh38
rs782030305 Chr.19: 8584977-8584977 on Build GRCh38
rs530244308 Chr.19: 8584915-8584915 on Build GRCh38
rs10405956 Chr.19: 8584995-8584995 on Build GRCh38
rs1027851957 Chr.19: 8584829-8584829 on Build GRCh38

View Full Product Details