The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006896.3 | NP_008827.2 | 1 - 2 | 510 | 131 |
GenBank mRNA | AF026397.1 | - | 1 - 2 | 510 | 131 |