The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017410.2 | NP_059106.2 | 1 - 2 | 850 | 143 |
GenBank mRNA | AF263466.2 | - | 1 - 2 | 807 | 143 |
AK024027.1 | - | 1 - 2 | 332 | 143 | |
BC002754.1 | - | 1 - 2 | 797 | 143 | |
BC090850.1 | - | 1 - 2 | 819 | 143 | |
BT009908.1 | - | 1 - 2 | 735 | 143 | |
BU158600.1 | - | 1 - 2 | 139 | 143 |