The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032864.3 | NP_116253.2 | 2 - 3 | 495 | 103 |
XM_011542315.2 | XP_011540617.1 | 2 - 3 | 495 | 103 | |
GenBank mRNA | AK027842.1 | - | 1 - 2 | 103 | |
AK092038.1 | - | 2 - 3 | 478 | 103 | |
BC105004.1 | - | 2 - 3 | 336 | 103 | |
BC105006.1 | - | 2 - 3 | 336 | 103 | |
DA055001.1 | - | 2 - 3 | 495 | 103 |