The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145294.4 | NP_660337.3 | 23 - 24 | 2946 | 107 |
XM_017023023.1 | XP_016878512.1 | 23 - 24 | 2964 | 107 | |
XM_017023024.1 | XP_016878513.1 | 5 - 6 | 577 | 107 | |
GenBank mRNA | AB067511.1 | - | 20 - 21 | 4975 | 107 |
AK093802.1 | - | 3 - 4 | 323 | 107 | |
AK126622.1 | - | 9 - 10 | 1585 | 107 | |
AK131510.1 | - | 6 - 7 | 870 | 107 |