The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021014.3 | NP_066294.1 | 1 - 2 | 126 | 84 |
XM_011543885.2 | XP_011542187.1 | 1 - 2 | 79 | 84 | |
GenBank mRNA | AK310348.1 | - | 1 - 2 | 68 | 84 |
AK310513.1 | - | 1 - 2 | 79 | 84 | |
BC005904.1 | - | 1 - 2 | 80 | 84 | |
BC103862.1 | - | 1 - 2 | 125 | 84 | |
DB444418.1 | - | 1 - 2 | 220 | 84 |