Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 88431057-88431320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA7
Entrez Gene ID
55812
Gene Name
spermatogenesis associated 7
Gene Aliases
HEL-S-296, HSD-3.1, HSD3, LCA3
UniGene
Hs.525518
Gene Chromosome Location
Chr.14: 88385638-88470350 on Build GRCh38
Transcripts
NM_001040428.3 NM_018418.4 XM_005267851.1 XM_005267852.1 XM_005267854.1 XM_005267855.1 XM_006720204.1 XM_006720205.1 XM_011536951.1 XM_011536952.1 XM_011536953.1 XM_017021452.1 XM_017021453.1 XM_017021454.1 XM_017021455.1 XM_017021456.1 XM_017021457.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2250311 c.1033A>G p.M345V Chr.14: 88431176-88431176 on Build GRCh38
2250313 c.1042T>C p.Y348H Chr.14: 88431185-88431185 on Build GRCh38
401504 c.1029-2A>T p.? Chr.14: 88431170-88431170 on Build GRCh38
699169 c.1066C>A p.R356S Chr.14: 88431209-88431209 on Build GRCh38
6141560 c.1040A>T p.Q347L Chr.14: 88431183-88431183 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs34349775 Chr.14: 88431267-88431268 on Build GRCh38
rs751818717 Chr.14: 88431208-88431208 on Build GRCh38
rs376148770 Chr.14: 88431257-88431257 on Build GRCh38
rs1039878203 Chr.14: 88431105-88431105 on Build GRCh38
rs749418197 Chr.14: 88431243-88431243 on Build GRCh38
rs780041447 Chr.14: 88431133-88431133 on Build GRCh38
rs188413736 Chr.14: 88431117-88431117 on Build GRCh38
rs772844634 Chr.14: 88431143-88431143 on Build GRCh38
rs763142647 Chr.14: 88431138-88431138 on Build GRCh38
rs570862555 Chr.14: 88431112-88431112 on Build GRCh38
rs878853385 Chr.14: 88431201-88431202 on Build GRCh38
rs752105058 Chr.14: 88431269-88431269 on Build GRCh38
rs780148709 Chr.14: 88431230-88431230 on Build GRCh38
rs151154007 Chr.14: 88431210-88431210 on Build GRCh38
rs764804249 Chr.14: 88431262-88431262 on Build GRCh38
rs763777521 Chr.14: 88431114-88431114 on Build GRCh38
rs778354076 Chr.14: 88431139-88431139 on Build GRCh38
rs753833055 Chr.14: 88431151-88431151 on Build GRCh38
rs771869237 Chr.14: 88431142-88431142 on Build GRCh38
rs759599666 Chr.14: 88431261-88431261 on Build GRCh38
rs777510079 Chr.14: 88431199-88431199 on Build GRCh38
rs775684460 Chr.14: 88431137-88431137 on Build GRCh38
rs375371982 Chr.14: 88431176-88431176 on Build GRCh38
rs766172057 Chr.14: 88431145-88431145 on Build GRCh38
rs752372338 Chr.14: 88431184-88431184 on Build GRCh38
rs770919276 Chr.14: 88431250-88431250 on Build GRCh38
rs769866084 Chr.14: 88431124-88431124 on Build GRCh38
rs769850758 Chr.14: 88431223-88431223 on Build GRCh38
rs937449352 Chr.14: 88431106-88431106 on Build GRCh38
rs547142371 Chr.14: 88431244-88431244 on Build GRCh38
rs758022550 Chr.14: 88431192-88431192 on Build GRCh38
rs1041663408 Chr.14: 88431280-88431283 on Build GRCh38
rs867220839 Chr.14: 88431189-88431189 on Build GRCh38
rs764902056 Chr.14: 88431178-88431178 on Build GRCh38
rs760541934 Chr.14: 88431144-88431144 on Build GRCh38
rs760322259 Chr.14: 88431248-88431248 on Build GRCh38
rs534836613 Chr.14: 88431278-88431278 on Build GRCh38
rs373199356 Chr.14: 88431247-88431247 on Build GRCh38
rs942936781 Chr.14: 88431091-88431091 on Build GRCh38
rs746439416 Chr.14: 88431220-88431220 on Build GRCh38
rs369170423 Chr.14: 88431209-88431209 on Build GRCh38

View Full Product Details