The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002909.4 | NP_002900.2 | 1 - 2 | 56 | 92 |
GenBank mRNA | BM769434.1 | - | 1 - 2 | 68 | 92 |