The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003106.3 | NP_003097.1 | 1 - 1 | 164 | 121 |
GenBank mRNA | AW163619.1 | - | 1 - 1 | 154 | 121 |
DA388996.1 | - | 1 - 1 | 162 | 121 |