The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301036.1 | NP_001287965.1 | 3 - 4 | 1285 | 129 |
NM_020698.3 | NP_065749.2 | 3 - 4 | 1266 | 129 | |
GenBank mRNA | AB032971.1 | - | 3 - 4 | 1195 | 129 |
BC040535.1 | - | 3 - 4 | 1252 | 129 |