The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168215.1 | NP_001161687.1 | 2 - 3 | 182 | 72 |
NM_153229.2 | NP_694961.2 | 1 - 2 | 147 | 72 | |
XM_006721730.3 | XP_006721793.1 | 1 - 2 | 147 | 72 | |
GenBank mRNA | AK090637.1 | - | 1 - 2 | 102 | 72 |
AY358204.1 | - | 2 - 3 | 182 | 72 | |
BC064507.1 | - | 1 - 2 | 75 | 72 | |
DB072318.1 | - | 1 - 2 | 148 | 73 |