The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321057.1 | NP_001307986.1 | 5 - 6 | 2136 | 118 |
NM_176810.2 | NP_789780.2 | 5 - 6 | 2136 | 118 | |
GenBank mRNA | AY154468.1 | - | 5 - 6 | 2111 | 118 |