Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3583008-3583281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLX4
Entrez Gene ID
84464
Gene Name
SLX4 structure-specific endonuclease subunit
Gene Aliases
BTBD12, FANCP, MUS312
UniGene
Hs.143681
Gene Chromosome Location
Chr.16: 3581181-3611598 on Build GRCh38
Transcripts
NM_032444.2 XM_011522715.2 XM_017023775.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4728187 c.5033delC p.P1678fs*37 Chr.16: 3583217-3583217 on Build GRCh38
5627479 c.5117C>T p.S1706F Chr.16: 3583133-3583133 on Build GRCh38
7390981 c.5146T>A p.S1716T Chr.16: 3583104-3583104 on Build GRCh38
703056 c.5101G>C p.E1701Q Chr.16: 3583149-3583149 on Build GRCh38
3888511 c.5042C>T p.S1681L Chr.16: 3583208-3583208 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374696176 Chr.16: 3583078-3583078 on Build GRCh38
rs139304111 Chr.16: 3583250-3583250 on Build GRCh38
rs377544881 Chr.16: 3583223-3583223 on Build GRCh38
rs529888376 Chr.16: 3583074-3583074 on Build GRCh38
rs754209356 Chr.16: 3583091-3583091 on Build GRCh38
rs774540571 Chr.16: 3583070-3583070 on Build GRCh38
rs771106417 Chr.16: 3583069-3583069 on Build GRCh38
rs757013389 Chr.16: 3583235-3583235 on Build GRCh38
rs184699331 Chr.16: 3583154-3583154 on Build GRCh38
rs771669205 Chr.16: 3583060-3583060 on Build GRCh38
rs1034939893 Chr.16: 3583094-3583094 on Build GRCh38
rs768122851 Chr.16: 3583080-3583080 on Build GRCh38
rs755815284 Chr.16: 3583117-3583117 on Build GRCh38
rs868774001 Chr.16: 3583206-3583206 on Build GRCh38
rs765960272 Chr.16: 3583190-3583190 on Build GRCh38
rs551385115 Chr.16: 3583178-3583178 on Build GRCh38
rs771178344 Chr.16: 3583143-3583143 on Build GRCh38
rs7196345 Chr.16: 3583221-3583221 on Build GRCh38
rs762561686 Chr.16: 3583057-3583058 on Build GRCh38
rs756047873 Chr.16: 3583215-3583215 on Build GRCh38
rs75182789 Chr.16: 3583104-3583104 on Build GRCh38
rs745341094 Chr.16: 3583044-3583044 on Build GRCh38
rs776096631 Chr.16: 3583167-3583167 on Build GRCh38
rs530123827 Chr.16: 3583208-3583208 on Build GRCh38
rs367566849 Chr.16: 3583174-3583174 on Build GRCh38
rs569861974 Chr.16: 3583109-3583109 on Build GRCh38
rs772278475 Chr.16: 3583165-3583165 on Build GRCh38
rs1022072026 Chr.16: 3583152-3583153 on Build GRCh38
rs760878902 Chr.16: 3583081-3583081 on Build GRCh38
rs202099433 Chr.16: 3583153-3583153 on Build GRCh38
rs766089444 Chr.16: 3583100-3583100 on Build GRCh38
rs1045464630 Chr.16: 3583207-3583207 on Build GRCh38
rs898554987 Chr.16: 3583041-3583041 on Build GRCh38
rs374109750 Chr.16: 3583065-3583065 on Build GRCh38
rs761226343 Chr.16: 3583169-3583169 on Build GRCh38
rs925871189 Chr.16: 3583140-3583140 on Build GRCh38
rs754498907 Chr.16: 3583106-3583106 on Build GRCh38
rs746120772 Chr.16: 3583251-3583251 on Build GRCh38
rs751219957 Chr.16: 3583194-3583194 on Build GRCh38
rs937014008 Chr.16: 3583213-3583213 on Build GRCh38
rs987462786 Chr.16: 3583182-3583182 on Build GRCh38
rs988571866 Chr.16: 3583171-3583171 on Build GRCh38
rs748115513 Chr.16: 3583111-3583111 on Build GRCh38
rs370334914 Chr.16: 3583090-3583090 on Build GRCh38
rs967813945 Chr.16: 3583157-3583157 on Build GRCh38
rs199592185 Chr.16: 3583210-3583210 on Build GRCh38
rs769636026 Chr.16: 3583058-3583058 on Build GRCh38
rs777602041 Chr.16: 3583141-3583141 on Build GRCh38
rs772475203 Chr.16: 3583253-3583253 on Build GRCh38
rs987938943 Chr.16: 3583115-3583115 on Build GRCh38
rs745883123 Chr.16: 3583162-3583162 on Build GRCh38
rs192310418 Chr.16: 3583040-3583040 on Build GRCh38
rs753651021 Chr.16: 3583234-3583234 on Build GRCh38
rs376024875 Chr.16: 3583256-3583256 on Build GRCh38
rs754908078 Chr.16: 3583046-3583046 on Build GRCh38
rs762144948 Chr.16: 3583181-3583181 on Build GRCh38
rs377607188 Chr.16: 3583047-3583047 on Build GRCh38
rs779206091 Chr.16: 3583243-3583243 on Build GRCh38
rs747126736 Chr.16: 3583258-3583258 on Build GRCh38

View Full Product Details