The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306154.1 | NP_001293083.1 | 1 - 2 | 523 | 123 |
NM_012073.4 | NP_036205.1 | 1 - 2 | 523 | 123 | |
GenBank mRNA | AK289353.1 | - | 1 - 2 | 121 | 123 |
AK290393.1 | - | 1 - 2 | 164 | 123 | |
AK293437.1 | - | 1 - 2 | 126 | 123 | |
AK301985.1 | - | 1 - 2 | 162 | 123 | |
AK302368.1 | - | 1 - 2 | 228 | 123 | |
AK303086.1 | - | 1 - 2 | 152 | 123 | |
BC002971.1 | - | 1 - 2 | 98 | 123 | |
BC006543.2 | - | 1 - 2 | 138 | 123 | |
BC035499.2 | - | 1 - 2 | 185 | 123 | |
BC040027.1 | - | 1 - 2 | 155 | 123 | |
BU858349.1 | - | 1 - 2 | 291 | 123 | |
D43950.2 | - | 1 - 2 | 140 | 123 | |
FJ224303.1 | - | 1 - 2 | 274 | 123 |