The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012081.5 | NP_036213.2 | 6 - 7 | 1214 | 146 |
XM_006714575.2 | XP_006714638.1 | 7 - 8 | 1054 | 146 | |
XM_017009239.1 | XP_016864728.1 | 6 - 7 | 992 | 146 | |
XM_017009240.1 | XP_016864729.1 | 7 - 8 | 1481 | 146 | |
XM_017009241.1 | XP_016864730.1 | 5 - 6 | 1309 | 146 | |
XM_017009242.1 | XP_016864731.1 | 6 - 7 | 922 | 146 | |
XM_017009243.1 | XP_016864732.1 | 6 - 7 | 1525 | 146 | |
GenBank mRNA | AB209343.1 | - | 3 - 4 | 503 | 146 |
BC028412.2 | - | 6 - 7 | 1216 | 146 |