Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITPRIP
Entrez Gene ID
85450
Gene Name
inositol 1,4,5-trisphosphate receptor interacting protein
Gene Aliases
DANGER, KIAA1754, bA127L20, bA127L20.2
Chromosome Location
Chr.10: 104309696 - 104338493 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033397.3 NP_203755.1 2 - 3 440 160
GenBank mRNA BC040536.1 - 1 - 2 100 160
BC070108.1 - 2 - 3 440 160

View Full Product Details