The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152729.2 | NP_689942.2 | 9 - 10 | 1019 | 74 |
GenBank mRNA | AK090918.1 | - | 6 - 7 | 1749 | 74 |
AK290408.1 | - | 9 - 10 | 969 | 74 | |
AK308337.1 | - | 9 - 10 | 1002 | 74 | |
AY203935.1 | - | 6 - 7 | 629 | 74 | |
BC015138.1 | - | 8 - 9 | 799 | 74 | |
BC108699.1 | - | 9 - 10 | 976 | 74 | |
BC136832.1 | - | 9 - 10 | 978 | 74 | |
BC136841.1 | - | 9 - 10 | 978 | 74 |