Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IGDCC3
Entrez Gene ID
9543
Gene Name
immunoglobulin superfamily DCC subclass member 3
Gene Aliases
HsT18880, PUNC
Chromosome Location
Chr.15: 65327127 - 65378040 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004884.3 NP_004875.2 11 - 12 2106 71
XM_011522241.1 XP_011520543.1 11 - 12 2048 71
XM_011522243.1 XP_011520545.1 10 - 11 1632 71
XM_011522244.1 XP_011520546.1 10 - 11 1621 71
GenBank mRNA AF063936.1 - 3 - 4 338 71
AK095529.1 - 10 - 11 1621 71
BC042054.1 - 11 - 12 2054 71
BC067107.1 - 11 - 12 2106 71
BX389424.1 - 1 - 2 43 71

View Full Product Details