Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IGDCC3
Entrez Gene ID
9543
Gene Name
immunoglobulin superfamily DCC subclass member 3
Gene Aliases
HsT18880, PUNC
Chromosome Location
Chr.15: 65327127 - 65378040 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004884.3 NP_004875.2 13 - 14 2461 125
XM_011522243.1 XP_011520545.1 12 - 13 1987 125
XM_011522244.1 XP_011520546.1 12 - 13 1976 125
GenBank mRNA AF063936.1 - 5 - 6 693 125
BC042054.1 - 13 - 14 2409 125
BC067107.1 - 13 - 14 2461 125
BX389424.1 - 3 - 4 398 125

View Full Product Details