The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022075.4 | NP_071358.1 | 10 - 11 | 1391 | 68 |
NM_181746.3 | NP_859530.1 | 10 - 11 | 1431 | 68 | |
XM_011509451.2 | XP_011507753.1 | 10 - 11 | 1094 | 68 | |
XM_011509452.2 | XP_011507754.1 | 10 - 11 | 1142 | 68 | |
GenBank mRNA | AF177338.2 | - | 10 - 11 | 1120 | 68 |
AK001105.1 | - | 9 - 10 | 1060 | 68 | |
AK290989.1 | - | 10 - 11 | 1067 | 68 | |
AY091458.1 | - | 10 - 11 | 1213 | 68 | |
BC001357.2 | - | 10 - 11 | 1086 | 68 | |
BC010032.2 | - | 10 - 11 | 1050 | 68 |